Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019353.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019353.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019353.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019352.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019352.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019352.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019362.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019361.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019363.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019368.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019344.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019344.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019364.mm.1 |
TC0200002398.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_001042487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_001042487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_001042488.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_001277186.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_001277187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
NM_146128.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019365.mm.1 |
TC0200002398.mm.1 |
MTA TC |