Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145970.1 | NP_666082.2 | 6 - 7 | 784 | 77 | |
GenBank mRNA | BC016188.1 | - | 6 - 7 | 784 | 4482793 | 77 |
BC027028.1 | - | 5 - 6 | 599 | 5363547 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |