Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017390.4 | NP_059086.3 | 1 - 2 | 168 | 77 | |
GenBank mRNA | AK040724.1 | - | 1 - 2 | 94 | 77 | |
AK136679.1 | - | 1 - 2 | 92 | 77 | ||
AK144285.1 | - | 1 - 2 | 93 | 77 | ||
AK144389.1 | - | 1 - 2 | 93 | 77 | ||
AK144429.1 | - | 1 - 2 | 93 | 77 | ||
AK144524.1 | - | 1 - 2 | 90 | 77 | ||
AK165704.1 | - | 1 - 2 | 93 | 77 | ||
AK165713.1 | - | 1 - 2 | 93 | 77 | ||
AK165727.1 | - | 1 - 2 | 90 | 77 | ||
AK165753.1 | - | 1 - 2 | 93 | 77 | ||
AK165825.1 | - | 1 - 2 | 93 | 77 | ||
BB871464.1 | - | 1 - 2 | 160 | 77 | ||
BC107275.1 | - | 1 - 2 | 93 | 40052650 | 77 | |
BC107276.1 | - | 1 - 2 | 93 | 77 | ||
DV056729.1 | - | 1 - 2 | 202 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |