Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029557.1 | NP_083833.1 | 2 - 3 | 247 | 100 | |
GenBank mRNA | AK002758.1 | - | 2 - 3 | 232 | 100 | |
AK089005.1 | - | 2 - 3 | 214 | 100 | ||
BC126953.1 | - | 1 - 2 | 371 | 100 | ||
BY203746.1 | - | 2 - 3 | 252 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |