Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029557.1 | NP_083833.1 | 4 - 5 | 398 | 67 | |
GenBank mRNA | AK002758.1 | - | 4 - 5 | 383 | 67 | |
AK089005.1 | - | 4 - 5 | 365 | 67 | ||
AK182728.1 | - | 1 - 2 | 88 | 67 | ||
BC126953.1 | - | 3 - 4 | 522 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |