Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252384.1 | NP_001239313.1 | 6 - 7 | 1072 | 84 | |
NM_001252395.1 | NP_001239324.1 | 6 - 7 | 1084 | 84 | ||
NM_001252396.1 | NP_001239325.1 | 4 - 5 | 739 | 84 | ||
NM_026071.3 | NP_080347.1 | 5 - 6 | 909 | 84 | ||
GenBank mRNA | AK005710.1 | - | 5 - 6 | 754 | 84 | |
AK172622.1 | - | 4 - 5 | 711 | 84 | ||
BC018167.1 | - | 5 - 6 | 715 | 3989263 | 84 | |
BC046767.1 | - | 5 - 6 | 759 | 5694117 | 84 | |
CF730205.1 | - | 4 - 5 | 550 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |