Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080643.2 | NP_542374.2 | 13 - 14 | 1858 | 79 | |
GenBank mRNA | AK129297.1 | - | 13 - 14 | 1896 | 79 | |
BC053083.1 | - | 13 - 14 | 1858 | 5704642 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |