Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010350.2 | NP_034480.2 | 10 - 11 | 2548 | 63 | |
GenBank mRNA | AK147390.1 | - | 9 - 10 | 2276 | 63 | |
AK165770.1 | - | 7 - 8 | 1403 | 63 | ||
BC137849.1 | - | 8 - 9 | 1938 | 8860868 | 63 | |
D10694.1 | - | 7 - 8 | 1775 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |