Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172948.3 | NP_766536.2 | 10 - 11 | 1908 | 61 | |
GenBank mRNA | AB116028.1 | - | 10 - 11 | 1295 | 61 | |
AB119127.1 | - | 10 - 11 | 1374 | 61 | ||
BC063084.1 | - | 9 - 10 | 1811 | 61 | ||
BC094604.1 | - | 10 - 11 | 1908 | 6822690 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |