Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175263.4 | NP_780472.3 | 7 - 8 | 793 | 70 | |
GenBank mRNA | AK019987.1 | - | 7 - 8 | 793 | 70 | |
BC025832.1 | - | 2 - 3 | 263 | 5125723 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |