Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195023.1 | NP_001181952.1 | 8 - 9 | 1014 | 68 | |
NM_199469.2 | NP_955763.1 | 8 - 9 | 1014 | 68 | ||
GenBank mRNA | AK129375.1 | - | 8 - 9 | 989 | 68 | |
AK141998.1 | - | 8 - 9 | 1013 | 68 | ||
AK149823.1 | - | 8 - 9 | 986 | 68 | ||
BC065156.1 | - | 8 - 9 | 982 | 5706754 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |