Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133223.4 | NP_573486.1 | 1 - 2 | 180 | 56 | |
GenBank mRNA | AB040819.1 | - | 1 - 2 | 47 | 56 | |
AK089930.1 | - | 1 - 2 | 181 | 56 | ||
AK150203.1 | - | 1 - 2 | 185 | 56 | ||
BC117028.1 | - | 1 - 2 | 178 | 40126347 | 56 | |
BC119041.1 | - | 1 - 2 | 178 | 8733790 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |