Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198423.3 | NP_940815.3 | 21 - 22 | 6213 | 72 | |
GenBank mRNA | AB257855.1 | - | 20 - 21 | 6215 | 72 | |
AK147250.1 | - | 21 - 22 | 6213 | 72 | ||
AK147400.1 | - | 13 - 14 | 3121 | 72 | ||
AK164229.1 | - | 4 - 5 | 454 | 72 | ||
BC060615.1 | - | 4 - 5 | 485 | 72 | ||
BC072602.1 | - | 10 - 11 | 1651 | 6830770 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |