Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198423.3 | NP_940815.3 | 11 - 12 | 4480 | 57 | |
GenBank mRNA | AB257855.1 | - | 10 - 11 | 4482 | 57 | |
AK147250.1 | - | 11 - 12 | 4480 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |