Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026197.3 | NP_080473.1 | 2 - 3 | 239 | 113 | |
GenBank mRNA | AK011780.1 | - | 2 - 3 | 240 | 113 | |
AK012739.1 | - | 2 - 3 | 240 | 113 | ||
AK013306.1 | - | 2 - 3 | 251 | 113 | ||
AK029045.1 | - | 2 - 3 | 223 | 113 | ||
AK145022.1 | - | 2 - 3 | 247 | 113 | ||
AK145662.1 | - | 2 - 3 | 239 | 113 | ||
AK168798.1 | - | 2 - 3 | 189 | 113 | ||
BC003467.1 | - | 2 - 3 | 210 | 113 | ||
BC022902.1 | - | 2 - 3 | 251 | 113 | ||
BC037452.1 | - | 2 - 3 | 188 | 113 | ||
BC049897.1 | - | 2 - 3 | 168 | 3979938 | 113 | |
BC054724.1 | - | 2 - 3 | 175 | 5695738 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |