Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247720 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247726 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247719 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247723 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247722 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247733 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247727 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247736 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247732 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247724 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247728 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247740 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247735 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247717 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247721 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247734 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247730 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247738 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P114528 |
BC094596.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P114528 |
NM_198632.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933810 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
443369 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
339547 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
721898 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510286 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
362148 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
646266 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
231715 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
430278 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895120 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
77395 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
724740 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57699 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700716 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579375 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134625 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867666 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
494576 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
86580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462781 |
AK050399.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462781 |
AK144479.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462781 |
NM_001033478.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
167359 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899970 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437970 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
114861 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
99976 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5911 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
13969 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168564 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P114528 |
BC094596.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P114528 |
NM_198632.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01852976 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06675795 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168564 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01853011 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06675836 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06675776 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P342837 |
AK220282.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P342837 |
AK051455.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P1130990 |
BC094596.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P1130990 |
NM_198632.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P111412 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06675776 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168564 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247728 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247740 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247730 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247732 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247719 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247734 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247733 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247736 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247720 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247727 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247724 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247723 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247738 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247735 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247717 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247726 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247721 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26247722 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247700 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247689 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247697 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247761 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247740 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247753 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247737 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07106161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06797871 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247681 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247776 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247680 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247702 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247694 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247685 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247690 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247720 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247723 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247711 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247732 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247767 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07104045 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247707 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247703 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07004633 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247683 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247730 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247738 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247705 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06926376 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247715 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247696 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07106149 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247686 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07270970 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247766 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06872643 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247774 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247714 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07071606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247764 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07438785 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247708 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247768 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06756663 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06800056 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07004640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247770 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26247757 |
- |
- |