Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021456.4 | NP_067431.2 | 7 - 8 | 956 | 101 | |
GenBank mRNA | AK136446.1 | - | 7 - 8 | 953 | 101 | |
AK136588.1 | - | 7 - 8 | 953 | 101 | ||
AK149446.1 | - | 7 - 8 | 953 | 101 | ||
AK162328.1 | - | 7 - 8 | 931 | 101 | ||
BC021150.1 | - | 7 - 8 | 975 | 4163576 | 101 | |
BC026897.1 | - | 7 - 8 | 936 | 4195247 | 101 | |
Y12887.1 | - | 7 - 8 | 942 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |