Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023311.3 | NP_075800.1 | 1 - 2 | 176 | 106 | |
GenBank mRNA | AK007671.1 | - | 1 - 2 | 136 | 106 | |
AK010089.1 | - | 1 - 2 | 136 | 106 | ||
AK151425.1 | - | 1 - 2 | 96 | 106 | ||
AK151461.1 | - | 1 - 2 | 96 | 106 | ||
AK152440.1 | - | 1 - 2 | 138 | 106 | ||
AK153123.1 | - | 1 - 2 | 135 | 106 | ||
AK160004.1 | - | 1 - 2 | 114 | 112 | ||
AK167526.1 | - | 1 - 2 | 136 | 100 | ||
AK167953.1 | - | 1 - 2 | 107 | 106 | ||
AK172133.1 | - | 1 - 2 | 114 | 106 | ||
BC003317.1 | - | 1 - 2 | 105 | 3155690 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |