Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175199.3 | NP_780408.1 | 3 - 4 | 350 | 58 | |
GenBank mRNA | AB093239.1 | - | 3 - 4 | 294 | 58 | |
AK006867.1 | - | 3 - 4 | 334 | 58 | ||
AK038491.1 | - | 3 - 4 | 322 | 58 | ||
AK087732.1 | - | 4 - 5 | 507 | 58 | ||
AK141925.1 | - | 3 - 4 | 351 | 58 | ||
AK165073.1 | - | 3 - 4 | 378 | 58 | ||
AY196789.1 | - | 3 - 4 | 349 | 58 | ||
BC030362.1 | - | 3 - 4 | 320 | 5369051 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |