Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175199.3 | NP_780408.1 | 9 - 10 | 1120 | 127 | |
GenBank mRNA | AB093239.1 | - | 9 - 10 | 1064 | 127 | |
AK038491.1 | - | 9 - 10 | 1092 | 127 | ||
AK087732.1 | - | 10 - 11 | 1278 | 127 | ||
AK141925.1 | - | 9 - 10 | 1121 | 127 | ||
AY196789.1 | - | 9 - 10 | 1119 | 127 | ||
BC030362.1 | - | 9 - 10 | 1090 | 5369051 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |