Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018817.2 | NP_061287.2 | 11 - 12 | 2188 | 73 | |
GenBank mRNA | AF088884.1 | - | 11 - 12 | 2168 | 73 | |
AK077878.1 | - | 11 - 12 | 2166 | 73 | ||
AK083488.1 | - | 11 - 12 | 2166 | 73 | ||
AK146767.1 | - | 11 - 12 | 2184 | 73 | ||
AK154020.1 | - | 11 - 12 | 2171 | 73 | ||
AK161570.1 | - | 10 - 11 | 3480 | 73 | ||
BC029078.1 | - | 11 - 12 | 2123 | 3992325 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |