Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010518.2 | NP_034648.2 | 2 - 3 | 1293 | 66 | |
GenBank mRNA | AK142108.1 | - | 2 - 3 | 1313 | 66 | |
AK161584.1 | - | 1 - 2 | 102 | 66 | ||
BC026805.1 | - | 2 - 3 | 733 | 66 | ||
BC029321.1 | - | 1 - 2 | 127 | 66 | ||
BC054812.1 | - | 2 - 3 | 703 | 6841070 | 66 | |
BC057447.1 | - | 2 - 3 | 1293 | 2648602 | 66 | |
L12447.1 | - | 2 - 3 | 954 | 66 | ||
X81583.1 | - | 2 - 3 | 630 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |