Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
TC0700000661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_001199015.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_001199015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_001199016.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_001199016.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_021291.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005607.mm.1 |
NM_021291.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
TC0700000661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_001199015.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_001199015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_001199016.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_001199016.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_021291.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005606.mm.1 |
NM_021291.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
TC0700000661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_001199015.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_001199015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_001199016.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_001199016.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_021291.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005608.mm.1 |
NM_021291.3 |
Refseq |