Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199015.1 | NP_001185944.1 | 9 - 10 | 1082 | 81 | |
NM_001199016.1 | NP_001185945.1 | 8 - 9 | 970 | 81 | ||
NM_021291.3 | NP_067266.1 | 8 - 9 | 1197 | 81 | ||
GenBank mRNA | AF192310.1 | - | 9 - 10 | 1082 | 81 | |
AJ249198.1 | - | 8 - 9 | 1197 | 81 | ||
AK142541.1 | - | 8 - 9 | 971 | 81 | ||
AK164714.1 | - | 7 - 8 | 1387 | 81 | ||
BC010746.1 | - | 9 - 10 | 1060 | 4236578 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |