Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026181.1 | NP_080457.1 | 12 - 13 | 1789 | 65 | |
GenBank mRNA | AK004868.1 | - | 12 - 13 | 1789 | 65 | |
AK014367.1 | - | 12 - 13 | 1768 | 65 | ||
AK154571.1 | - | 12 - 13 | 1770 | 65 | ||
BC026394.1 | - | 6 - 7 | 955 | 65 | ||
BC100577.1 | - | 12 - 13 | 1764 | 65 | ||
BC125584.1 | - | 12 - 13 | 1780 | 40130099 | 65 | |
BC132155.1 | - | 12 - 13 | 1780 | 40130432 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |