Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027696.3 | NP_081972.2 | 3 - 4 | 303 | 122 | |
GenBank mRNA | AY769435.1 | - | 2 - 3 | 95 | 122 | |
BC043930.1 | - | 3 - 4 | 228 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |