Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039081.2 | NP_001034170.1 | 7 - 8 | 724 | 127 | |
NM_001286221.1 | NP_001273150.1 | 9 - 10 | 855 | 127 | ||
NM_001286222.1 | NP_001273151.1 | 9 - 10 | 909 | 127 | ||
NM_001286223.1 | NP_001273152.1 | 7 - 8 | 705 | 127 | ||
NM_027696.3 | NP_081972.2 | 9 - 10 | 912 | 127 | ||
GenBank mRNA | AK016915.1 | - | 1 - 2 | 154 | 127 | |
AK129405.1 | - | 9 - 10 | 832 | 127 | ||
AY769435.1 | - | 8 - 9 | 704 | 127 | ||
AY769436.1 | - | 7 - 8 | 627 | 127 | ||
AY769437.1 | - | 7 - 8 | 677 | 127 | ||
AY769438.1 | - | 9 - 10 | 777 | 127 | ||
BC108273.1 | - | 7 - 8 | 691 | 30080243 | 127 | |
BC130227.1 | - | 7 - 8 | 676 | 40141863 | 127 | |
BQ562973.1 | - | 4 - 5 | 302 | 127 | ||
CN538186.1 | - | 4 - 5 | 325 | 127 | ||
CN701986.1 | - | 1 - 2 | 78 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |