Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081264.1 | NP_001074733.1 | 11 - 12 | 1185 | 162 | |
GenBank mRNA | AK157903.1 | - | 11 - 12 | 1147 | 162 | |
AK171887.1 | - | 11 - 12 | 1138 | 162 | ||
BC050854.1 | - | 11 - 12 | 1150 | 6332345 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |