Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081264.1 | NP_001074733.1 | 12 - 13 | 1262 | 144 | |
GenBank mRNA | AK157903.1 | - | 12 - 13 | 1224 | 144 | |
AK171887.1 | - | 12 - 13 | 1215 | 144 | ||
BC044875.1 | - | 2 - 3 | 97 | 144 | ||
BC050854.1 | - | 12 - 13 | 1227 | 6332345 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |