Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145844 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145841 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145840 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145843 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955687 |
BC050854.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955687 |
BC080867.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955687 |
AK171887.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955687 |
BC044875.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955687 |
AK157903.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955691 |
NM_001081264.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
348474 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404564 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250101 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593707 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211415 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
509681 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
AK048279.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
AK155021.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
AK165669.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
AK166648.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
AK170781.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911768 |
NM_009906.5 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931292 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
35398 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401789 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
17725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
730081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
228003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829755 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198158 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556649 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
107209 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579380 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
926649 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
670776 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
119353 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
599908 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
79907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83875 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81941 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137744 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169784 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144867 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955687 |
BC050854.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955687 |
BC080867.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955687 |
AK171887.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955687 |
BC044875.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955687 |
AK157903.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955691 |
NM_001081264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473209 |
BC050854.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473209 |
AK171887.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473209 |
BC044875.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473209 |
AK157903.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P473209 |
NM_001081264.1 |
mouse_transcriptome |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145841 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145840 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145844 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23145843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23145898 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03490172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145846 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145848 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145822 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145980 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03728330 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145836 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145791 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145795 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03425166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145828 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145985 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145854 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145984 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145900 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145875 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145878 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146072 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145794 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03374837 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145874 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145826 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145905 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145800 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145841 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145863 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145817 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145856 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145884 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145788 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145786 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145876 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145824 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03378505 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146068 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146067 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145986 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145981 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145886 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145812 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145885 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145989 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145879 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03219683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145872 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04047940 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145860 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145857 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145865 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145976 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146074 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145977 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145815 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145880 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03971957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145889 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145909 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145902 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145809 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145866 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145819 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145898 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145975 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145971 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145969 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145850 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03832354 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145845 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145870 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03582117 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145806 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145832 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145861 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145818 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145979 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145858 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145978 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03582112 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145793 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145798 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145904 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754849 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23145868 |
- |
- |