Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160038.1 | NP_001153510.1 | 14 - 15 | 1892 | 152 | |
NM_001160039.1 | NP_001153511.1 | 14 - 15 | 1734 | 152 | ||
NM_001160040.1 | NP_001153512.1 | 13 - 14 | 1788 | 152 | ||
NM_145518.2 | NP_663493.2 | 13 - 14 | 1630 | 152 | ||
GenBank mRNA | AK034597.1 | - | 13 - 14 | 1699 | 152 | |
AK036926.1 | - | 13 - 14 | 1495 | 152 | ||
AK142711.1 | - | 13 - 14 | 1510 | 152 | ||
AK167705.1 | - | 13 - 14 | 1478 | 152 | ||
BC006660.1 | - | 13 - 14 | 1517 | 3501043 | 152 | |
BC015300.1 | - | 13 - 14 | 1450 | 4236882 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |