Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160038.1 | NP_001153510.1 | 10 - 11 | 1376 | 152 | |
NM_001160039.1 | NP_001153511.1 | 10 - 11 | 1218 | 152 | ||
NM_001160040.1 | NP_001153512.1 | 9 - 10 | 1272 | 152 | ||
NM_145518.2 | NP_663493.2 | 9 - 10 | 1114 | 152 | ||
GenBank mRNA | AK034597.1 | - | 9 - 10 | 1183 | 152 | |
AK036926.1 | - | 9 - 10 | 979 | 152 | ||
AK078590.1 | - | 9 - 10 | 961 | 152 | ||
AK142711.1 | - | 9 - 10 | 994 | 152 | ||
AK146814.1 | - | 9 - 10 | 1132 | 152 | ||
AK166604.1 | - | 10 - 11 | 1065 | 152 | ||
AK167705.1 | - | 9 - 10 | 962 | 152 | ||
BC006660.1 | - | 9 - 10 | 1001 | 3501043 | 152 | |
BC015300.1 | - | 9 - 10 | 934 | 4236882 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |