Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081436.2 | NP_001074905.1 | 4 - 5 | 403 | 58 | |
NM_001114609.1 | NP_001108081.1 | 4 - 5 | 494 | 58 | ||
GenBank mRNA | AK049468.1 | - | 4 - 5 | 391 | 58 | |
BC010584.1 | - | 4 - 5 | 562 | 58 | ||
BQ030826.1 | - | 1 - 2 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |