Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008384.2 | NP_032410.2 | 4 - 5 | 808 | 76 | |
GenBank mRNA | AK078382.1 | - | 4 - 5 | 696 | 76 | |
BC025072.1 | - | 4 - 5 | 553 | 4456812 | 76 | |
U27295.1 | - | 4 - 5 | 657 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |