Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153556.1 | NP_705784.1 | 11 - 12 | 2561 | 83 | |
GenBank mRNA | BC028939.1 | - | 11 - 12 | 2561 | 5364251 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |