Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153556.1 | NP_705784.1 | 1 - 2 | 118 | 66 | |
GenBank mRNA | AK045010.1 | - | 1 - 2 | 89 | 66 | |
BC028939.1 | - | 1 - 2 | 118 | 5364251 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |