Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153556.1 | NP_705784.1 | 7 - 8 | 956 | 110 | |
GenBank mRNA | AK140689.1 | - | 8 - 9 | 1021 | 110 | |
BC028939.1 | - | 7 - 8 | 956 | 5364251 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |