Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028599.2 | NP_082875.1 | 2 - 3 | 231 | 74 | |
GenBank mRNA | AK010772.1 | - | 2 - 3 | 231 | 74 | |
AK152412.1 | - | 2 - 3 | 232 | 74 | ||
AK163221.1 | - | 2 - 3 | 205 | 74 | ||
AK168032.1 | - | 2 - 3 | 225 | 74 | ||
AK168163.1 | - | 2 - 3 | 233 | 74 | ||
AK169932.1 | - | 2 - 3 | 229 | 74 | ||
BC085177.1 | - | 2 - 3 | 266 | 74 | ||
BC117880.2 | - | 2 - 3 | 261 | 40096010 | 74 | |
BC117881.1 | - | 2 - 3 | 258 | 40096014 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |