Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027407.3 | NP_081683.3 | 2 - 3 | 456 | 79 | |
GenBank mRNA | AK029991.1 | - | 2 - 3 | 228 | 79 | |
AK158859.1 | - | 2 - 3 | 292 | 79 | ||
AK165131.1 | - | 2 - 3 | 282 | 79 | ||
BC079669.1 | - | 2 - 3 | 456 | 30355976 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |