Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172733.1 | NP_766321.1 | 6 - 7 | 664 | 82 | |
GenBank mRNA | AK012558.1 | - | 6 - 7 | 700 | 82 | |
AK034347.1 | - | 666 | 82 | |||
AK075963.1 | - | 6 - 7 | 689 | 82 | ||
AK083298.1 | - | 6 - 7 | 664 | 82 | ||
AK145820.1 | - | 6 - 7 | 700 | 82 | ||
AK150273.1 | - | 6 - 7 | 690 | 82 | ||
AK150712.1 | - | 6 - 7 | 660 | 82 | ||
AK152462.1 | - | 6 - 7 | 668 | 82 | ||
AK168204.1 | - | 6 - 7 | 700 | 82 | ||
AK169418.1 | - | 6 - 7 | 684 | 82 | ||
AK179075.1 | - | 2 - 3 | 170 | 82 | ||
AK199271.1 | - | 1 - 2 | 31 | 82 | ||
BC016218.1 | - | 6 - 7 | 670 | 4484163 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |