Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177192.3 | NP_796166.2 | 16 - 17 | 3439 | 68 | |
GenBank mRNA | AK135015.1 | - | 16 - 17 | 3322 | 68 | |
CN537548.1 | - | 4 - 5 | 436 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |