Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023617.2 | NP_076106.2 | 15 - 16 | 1735 | 99 | |
GenBank mRNA | AF172276.2 | - | 15 - 16 | 99 | ||
AK004700.1 | - | 4 - 5 | 531 | 99 | ||
AK030109.1 | - | 11 - 12 | 1275 | 99 | ||
AK149434.1 | - | 15 - 16 | 1696 | 99 | ||
AY347569.1 | - | 15 - 16 | 99 | |||
BC049853.1 | - | 15 - 16 | 1687 | 99 | ||
BC138876.1 | - | 15 - 16 | 8861898 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |