Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113367.1 | NP_001106838.1 | 5 - 6 | 592 | 75 | |
NM_029267.3 | NP_083543.2 | 5 - 6 | 537 | 75 | ||
GenBank mRNA | AF272859.1 | - | 5 - 6 | 400 | 75 | |
AK016403.1 | - | 5 - 6 | 592 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |