Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008514.4 | NP_032540.2 | 6 - 7 | 1590 | 127 | |
GenBank mRNA | AF074265.1 | - | 6 - 7 | 1459 | 127 | |
BC060704.1 | - | 6 - 7 | 1590 | 6409058 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |