Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stradb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172656.5 | NP_766244.4 | 6 - 7 | 797 | 123 | |
GenBank mRNA | AB057666.1 | - | 6 - 7 | 564 | 123 | |
AB057667.1 | - | 6 - 7 | 564 | 123 | ||
AK046790.1 | - | 770 | 123 | |||
AK080096.1 | - | 6 - 7 | 761 | 123 | ||
AK086974.1 | - | 6 - 7 | 728 | 123 | ||
AK140272.1 | - | 6 - 7 | 847 | 123 | ||
AK146079.1 | - | 6 - 7 | 610 | 123 | ||
BC037003.1 | - | 6 - 7 | 706 | 123 | ||
BC055814.1 | - | 7 - 8 | 937 | 123 | ||
BC060956.1 | - | 6 - 7 | 654 | 30316511 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |