Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033373.2 | NP_001028545.2 | 2 - 3 | 331 | 86 | |
GenBank mRNA | BC147314.1 | - | 2 - 3 | 331 | 9056364 | 86 |
BC147315.1 | - | 2 - 3 | 331 | 9056370 | 86 | |
BC147758.1 | - | 2 - 3 | 331 | 9087483 | 86 | |
BC147763.1 | - | 2 - 3 | 331 | 9087488 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |