Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164682.1 | NP_001158154.1 | 6 - 7 | 1114 | 67 | |
NM_145143.3 | NP_660125.2 | 5 - 6 | 477 | 67 | ||
GenBank mRNA | AJ748820.1 | - | 4 - 5 | 358 | 67 | |
AK044600.1 | - | 5 - 6 | 413 | 67 | ||
AK044682.1 | - | 6 - 7 | 1113 | 67 | ||
AK088771.1 | - | 166 | 67 | |||
AK138845.1 | - | 3 - 4 | 474 | 67 | ||
BC058776.1 | - | 6 - 7 | 490 | 67 | ||
BC061694.1 | - | 5 - 6 | 394 | 6468172 | 67 | |
BI737565.1 | - | 5 - 6 | 478 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |