Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161798.1 | NP_001155270.1 | 4 - 5 | 730 | 79 | |
NM_010840.3 | NP_034970.2 | 4 - 5 | 653 | 79 | ||
NR_027809.1 | 4 - 5 | 730 | 79 | |||
GenBank mRNA | AK030192.1 | - | 4 - 5 | 648 | 79 | |
AK149795.1 | - | 4 - 5 | 716 | 79 | ||
AK150159.1 | - | 4 - 5 | 731 | 79 | ||
AK219632.1 | - | 1 - 2 | 41 | 79 | ||
BC051017.1 | - | 4 - 5 | 632 | 6308248 | 79 | |
BC052466.1 | - | 4 - 5 | 697 | 6834886 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |