Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025365.2 | NP_001020536.2 | 5 - 6 | 1088 | 89 | |
GenBank mRNA | AK133976.1 | - | 7 - 8 | 1240 | 89 | |
AK170224.1 | - | 5 - 6 | 881 | 89 | ||
BC017525.1 | - | 5 - 6 | 831 | 4910951 | 89 | |
BC024069.1 | - | 5 - 6 | 870 | 5097088 | 89 | |
FJ618908.1 | - | 5 - 6 | 1088 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |