Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037737.2 | NP_001032826.1 | 11 - 12 | 1175 | 62 | |
NM_009709.4 | NP_033839.2 | 10 - 11 | 1130 | 62 | ||
GenBank mRNA | AK028546.1 | - | 10 - 11 | 1117 | 62 | |
AK040475.1 | - | 11 - 12 | 1177 | 62 | ||
AK141362.1 | - | 11 - 12 | 1046 | 62 | ||
AK145415.1 | - | 10 - 11 | 1085 | 62 | ||
AK152473.1 | - | 11 - 12 | 1157 | 62 | ||
AK153355.1 | - | 11 - 12 | 1157 | 62 | ||
AK220458.1 | - | 7 - 8 | 1306 | 62 | ||
BC012870.1 | - | 11 - 12 | 1137 | 3964932 | 62 | |
U10325.1 | - | 11 - 12 | 1039 | 62 | ||
U14333.1 | - | 10 - 11 | 968 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |